Invitrogen
Manufacturer Code:PA551754
Catalog # PIPA551754
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ALG13 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Asparagine-linked glycosylation 13 homolog; Glycosyltransferase 28 domain-containing protein 1; hematopoietic stem/progenitor cells protein MDS031; N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase; Putative bifunctional UDP-N-acetylglucosamine transferase and deubiquitinase ALG13; Putative bifunctional UDP-N-acetylglucosamine transferase and deubiquitinase ALG13 Asparagine-linked glycosylation 13 homolog Glycosyltransferase 28 domain-containing protein 1 UDP-N-acetylglucosamine transferase subunit ALG13 homolog ALG13 CXorf45 GLT28D1; tudor domain containing 13; UDP-N-acetylglucosamine transferase subunit ALG13 homolog
Gene Aliases: ALG13; CDG1S; CXorf45; EIEE36; GLT28D1; MDS031; TDRD13; YGL047W
UniProt ID: (Human) B1AKD6
Entrez Gene ID: (Human) 79868
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.