Proteintech
Manufacturer Code:208101AP150UL
Catalog # 50-173-3102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ALG13 Fusion Protein Ag14763 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALG13 CXorf45 GLT28D1 MDS031 YGL047W; Asparagine-linked glycosylation 13 homolog; glycosyltransferase 28 domain containing 1; Glycosyltransferase 28 domain-containing protein 1; hematopoietic stem/progenitor cells protein MDS031; N-acetylglucosaminyldiphosphodolichol N-acetylglucosaminyltransferase; Putative bifunctional UDP-N-acetylglucosamine transferase and deubiquitinase ALG13; tudor domain containing 13; UDP-N-acetylglucosamine transferase subunit ALG13 homolog
Gene Aliases: 2810046O15Rik; 4833435D08Rik; ALG13; CDG1S; CXorf45; EIEE36; GLT28D1; MDS031; OTTMUSG00000018917; TDRD13; YGL047W
UniProt ID: (Human) B1AKD6, (Rat) Q5I0K7, (Mouse) Q9D8C3
Entrez Gene ID: (Human) 79868, (Rat) 300284, (Mouse) 67574
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.