Proteintech
Manufacturer Code:270851AP150UL
Catalog # 50-173-5558
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ALG11 Fusion Protein Ag21123 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALG11 GT8 KIAA0266 UTP14C; asparagine-linked glycosylation 11 homolog (S. cerevisiae, alpha-1,2-mannosyltransferase); asparagine-linked glycosylation 11 homolog (yeast, alpha-1,2-mannosyltransferase); asparagine-linked glycosylation 11, alpha-1,2-mannosyltransferase homolog; Asparagine-linked glycosylation protein 11 homolog; defective mannosyltransferase IV(V); GDP-Man:Man(3)GlcNAc(2)-PP-Dol alpha-1,2-mannosyltransferase; GDP-Man:Man(3)GlcNAc(2)-PP-dolichol alpha-1,2-mannosyltransferase; Glycolipid 2-alpha-mannosyltransferase; mannosyltransferase IV(V)
Gene Aliases: AI849156; ALG11; AW492253; B230397C21; CDG1P; GT8; Mmat-4(5); RGD1564725
UniProt ID: (Human) Q2TAA5, (Mouse) Q3TZM9
Entrez Gene ID: (Human) 440138, (Mouse) 207958, (Rat) 361174
Molecular Function:
carbohydrate phosphatase
glycosyltransferase
hydrolase
nucleotidyltransferase
phosphatase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.