Invitrogen
Manufacturer Code:PA527919
Catalog # PIPA527919
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 59 and 350 of Human ALG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: asparagine-linked glycosylation 1 homolog (yeast, beta-1,4-mannosyltransferase); asparagine-linked glycosylation 1, beta-1,4-mannosyltransferase homolog; Asparagine-linked glycosylation protein 1 homolog; beta-1,4 mannosyltransferase; Beta-1,4-mannosyltransferase; CDG1K HMAT1 HMT-1 HMT1 MT-1 Mat-1 hMat-1 GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase GDP-mannose-dolichol diphosphochitobiose mannosyltransferase asparagine-linked glycosylation 1 homolog (yeast beta-14-mannosyltransferase) asparagine-linked glycosylation protein 1 homolog beta-14 mannosy; Chitobiosyldiphosphodolichol beta-mannosyltransferase; GDP-Man:GlcNAc2-PP-dolichol mannosyltransferase; GDP-mannose-dolichol diphosphochitobiose mannosyltransferase; Mannosyltransferase-1; MT-1
Gene Aliases: ALG1; CDG1K; hMat-1; HMAT1; HMT-1; HMT1; Mat-1; MT-1; PSEC0061; UNQ861/PRO1870
UniProt ID: (Human) Q9BT22
Entrez Gene ID: (Human) 56052
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.