Abnova
Manufacturer Code:PAB2569
Catalog # 89-127-311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human ALDOA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ALDOA.
For Research Use Only
Protein Aliases: ALDA; aldolase A, fructose-bisphosphate; epididymis secretory sperm binding protein Li 87p; fructose-1,6-bisphosphate triosephosphate-lyase; Fructose-bisphosphate aldolase A; Lung cancer antigen NY-LU-1; MGC10942; MGC17716; MGC17767; Muscle-type aldolase
Gene Aliases: ALDA; ALDOA; GSD12; HEL-S-87p
UniProt ID: (Human) P04075
Entrez Gene ID: (Human) 226
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.