Abnova
Manufacturer Code:PAB11351
Catalog # 89-115-035
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rabbit |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Native purified ALDOA from rabbit muscle. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with deionized water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against native ALDOA.
For Research Use Only
Protein Aliases: aldolase A; aldolase A, fructose-bisphosphate; Fructose-bisphosphate aldolase A; Muscle-type aldolase; unnamed protein product
Gene Aliases: ALDOA
UniProt ID: (Rabbit) P00883
Entrez Gene ID: (Rabbit) 100009055
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.