Invitrogen
Manufacturer Code:PA5112089
Catalog # PIPA5112089
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to the center region of the Human ALDH9A1/ALDH9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4-trimethylaminobutyraldehyde dehydrogenase; 4-trimethylaminobutyraldehyde dehydrogenase 4-trimethylaminobutyraldehyde dehydrogenase N-terminally processed AA139417 Abaldh aldehyde dehydrogenase (NAD+) aldehyde dehydrogenase 9 family member A1 aldehyde dehydrogenase 9 family member A1 aldehyde dehydrogenase 9 subfamily A1 aldehyde dehydrogenase 9A1 Aldehyde dehydrogenase E3 isozyme Aldehyde dehydrogenase family 9 member A1 aldehyde dehydrogenase family 9 subfamily A1 ALDH4 ALDH7 ALDH9 ALDH9A1 E3 ESTM40 Gamma-aminobutyraldehyde dehydrogenase gamma-trimethylaminobutyraldehyde dehydrogenase R-aminobutyraldehyde dehydrogenase TMABADH TMABA-DH; 4-trimethylaminobutyraldehyde dehydrogenase, N-terminally processed; aldehyde dehydrogenase (NAD+); aldehyde dehydrogenase 9 family, member A1; Aldehyde dehydrogenase E3 isozyme; Aldehyde dehydrogenase family 9 member A1; Gamma-aminobutyraldehyde dehydrogenase; R-aminobutyraldehyde dehydrogenase; TMABA-DH
Gene Aliases: ALDH4; ALDH7; ALDH9; ALDH9A1; E3; TMABADH
UniProt ID: (Human) P49189
Entrez Gene ID: (Human) 223
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.