Novus Biologicals
Manufacturer Code:NBP289927100UL
Catalog # NB021320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 105 |
Immunogen | Recombinant Human ALDH7A1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 26g turgor protein homolog; aldehyde dehydrogenase 7 family member A1 Aldehyde dehydrogenase family 7 member A126g turgor protein homolog Alpha-AASA dehydrogenase ATQ1Antiquitin-1 Betaine aldehyde dehydrogenase Delta1-piperideine-6-carboxylate dehydrogenase EC 1.2.1 EC 1.2.1.3 EC 1.2.1.31 EC 1.2.1.8 EPDalpha-aminoadipic semialdehyde dehydrogenase FLJ11738 FLJ92814 P6c dehydrogenase PDEantiquitin-1; aldehyde dehydrogenase 7 family, member A1; Aldehyde dehydrogenase family 7 member A1; Alpha-AASA dehydrogenase; Alpha-aminoadipic semialdehyde dehydrogenase; Antiquitin-1; Betaine aldehyde dehydrogenase; Delta1-piperideine-6-carboxylate dehydrogenase; P6c dehydrogenase
Gene Aliases: ALDH7A1; ATQ1; EPD; PDE
UniProt ID: (Human) P49419
Entrez Gene ID: (Human) 501
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.