Invitrogen
Manufacturer Code:PA585188
Catalog # PIPA585188
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human ALDH7A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 26g turgor protein homolog; aldehyde dehydrogenase 7 family, member A1; Aldehyde dehydrogenase family 7 member A1; Alpha-AASA dehydrogenase; Alpha-aminoadipic semialdehyde dehydrogenase; Antiquitin-1; ATQ1 antibody EPD antibody FLJ11738 antibody FLJ92814 antibody PDE antibody ALDH7A1 antibody antiquitin-1 antibody betaine aldehyde dehydrogenase antibody P6c dehydrogenase antibody alpha-aminoadipic semialdehyde dehydrogenase antibody alpha-AASA dehydrogenase antibody delta1-piperideine-6-carboxylate dehydrogenase antibody 26g turgor protein homolog antibody aldehyde dehydrogenase 7 family member A1 antibody; Betaine aldehyde dehydrogenase; Delta1-piperideine-6-carboxylate dehydrogenase; delta1-piperideine-6-carboxylate dehydrogenease; P6c dehydrogenase
Gene Aliases: Ald7a1; ALDH7A1; ATQ1; D18Wsu181e; EPD; PDE
UniProt ID: (Human) P49419, (Mouse) Q9DBF1
Entrez Gene ID: (Human) 501, (Mouse) 110695
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.