Invitrogen
Manufacturer Code:MA524714
Catalog # PIMA524714
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 147CT8.3.4 |
Immunogen | human ALDH6A1 recombit protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: aldehyde dehydrogenase 6 family, member A1; Aldehyde dehydrogenase family 6 member A1; malonate-semialdehyde dehydrogenase; Methylmalonate-semialdehyde dehydrogenase &lsqbacylating] mitochondrial MMSDH Malonate-semialdehyde dehydrogenase &lsqbacylating] Aldehyde dehydrogenase family 6 member A1 ALDH6A1 MMSDH; Methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial; mitochondrial acylating methylmalonate-semialdehyde dehydrogenase; MMSDH; testicular tissue protein Li 122
Gene Aliases: ALDH6A1; MMSADHA; MMSDH
UniProt ID: (Human) Q9UKM8
Entrez Gene ID: (Human) 4329
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.