Invitrogen
Manufacturer Code:PA519395
Catalog # PIPA519395
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (SRSSFRGDTN) corresponding to the internal amino acids of ALDH6A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: aldehyde dehydrogenase 6 family, member A1; Aldehyde dehydrogenase family 6 member A1; aldehyde dehydrogenase family 6, subfamily A1; ALDH6A1 aldehyde dehydrogenase 6 family member A1 MGC40271 MMSADHA MMSDH aldehyde dehydrogenase 6A1 mitochondrial acylating methylmalonate-semialdehyde dehydrogenase; malonate-semialdehyde dehydrogenase; methylmalonate semialdehyde dehydrogenase; Methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial; mitochondrial acylating methylmalonate-semialdehyde dehydrogenase; MMSDH; testicular tissue protein Li 122
Gene Aliases: 1110038I05Rik; AI314632; ALDH6A1; MMSADHA; MMSDH
UniProt ID: (Human) Q9UKM8, (Rat) Q02253, (Mouse) Q9EQ20
Entrez Gene ID: (Human) 4329, (Rat) 81708, (Mouse) 104776
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.