Novus Biologicals
Manufacturer Code:NBP152101
Catalog # NBP152101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SRSSFRGDTN corresponding to internal region according to NP_005580.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: aldehyde dehydrogenase 6 family member A1 Aldehyde dehydrogenase family 6 member A1 EC 1.2.1.18 EC 1.2.1.27 malonate-semialdehyde dehydrogenase Malonate-semialdehyde dehydrogenase [acylating] methylmalonate-semialdehyde dehydrogenase [acylating] mitochondrial MGC40271 MMSADHA MMSDHmitochondrial acylating methylmalonate-semialdehyde dehydrogenase; aldehyde dehydrogenase 6 family, member A1; Aldehyde dehydrogenase family 6 member A1; malonate-semialdehyde dehydrogenase; Methylmalonate-semialdehyde dehydrogenase [acylating], mitochondrial; mitochondrial acylating methylmalonate-semialdehyde dehydrogenase; MMSDH; testicular tissue protein Li 122
Gene Aliases: ALDH6A1; MMSADHA; MMSDH
UniProt ID: (Human) Q9UKM8
Entrez Gene ID: (Human) 4329
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.