OriGene
Manufacturer Code:TA812719
Catalog # 50-168-4662
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-503 of human ALDH4A1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: aldehyde dehydrogenase 4 family, member A1; Aldehyde dehydrogenase family 4 member A1; ALDH4 P5CD P5CDh; Delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial; L-glutamate gamma-semialdehyde dehydrogenase; mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase; P5C dehydrogenase
Gene Aliases: ALDH4; ALDH4A1; P5CD; P5CDH
UniProt ID: (Human) P30038
Entrez Gene ID: (Dog) 612452, (Human) 8659, (Rat) 641316
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.