Invitrogen
Manufacturer Code:MA529027
Catalog # PIMA529027
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2 |
Immunogen | Recombit Human ALDH4A1 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: aldehyde dehydrogenase 4 family, member A1; Aldehyde dehydrogenase family 4 member A1; Aldehyde dehydrogenase family 4 member A1 Delta-1-pyrroline-5-carboxylate dehydrogenase mitochondrial Short&equalsP5C dehydrogenase ALDH4A1 ALDH4 P5CDH; Delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial; L-glutamate gamma-semialdehyde dehydrogenase; mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase; P5C dehydrogenase
Gene Aliases: ALDH4; ALDH4A1; P5CD; P5CDH
UniProt ID: (Human) P30038
Entrez Gene ID: (Human) 8659
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.