Invitrogen
Manufacturer Code:PA542757
Catalog # PIPA542757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human ALDH4A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aldehyde dehydrogenase 4 family member A1 ALDH4 P5CD P5CDh P5CDhL P5CDhS; aldehyde dehydrogenase 4 family, member A1; aldehyde dehydrogenase 4A1; Aldehyde dehydrogenase family 4 member A1; Delta-1-pyrroline-5-carboxylate dehydrogenase, mitochondrial; L-glutamate gamma-semialdehyde dehydrogenase; mitochondrial delta-1-pyrroline 5-carboxylate dehydrogenase; P5C dehydrogenase
Gene Aliases: A930035F14Rik; Ahd-1; Ahd1; ALDH4; ALDH4A1; Aldh5a1; E330022C09; P5CD; P5CDH; P5cdhl; P5cdhs; Ssdh1; zgc:63592
UniProt ID: (Bovine) A7YWE4, (Human) P30038, (Mouse) Q8CHT0, (Zebrafish) Q7SY23
Entrez Gene ID: (Dog) 612452, (Horse) 100147246, (Rabbit) 100346729, (Pig) 100820830, (Bovine) 100126042, (Human) 8659, (Rat) 641316, (Mouse) 212647, (Zebrafish) 394133
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.