Novus Biologicals
Manufacturer Code:NBP286975
Catalog # NB034597
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of ALDH3B2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetaldehyde dehydrogenase 8; acetaldehyde dehydrogenase 8 aldehyde dehydrogenase 3 family member B2 Aldehyde dehydrogenase 8 ALDH8aldehyde dehydrogenase family 3 member B2 EC 1.2.1 EC 1.2.1.5; aldehyde dehydrogenase 3 family, member B2; aldehyde dehydrogenase 8
Gene Aliases: ALDH8
Entrez Gene ID: (Human) 222
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.