Proteintech
Manufacturer Code:157461AP150UL
Catalog # 50-173-0448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ALDH3B2 Fusion Protein Ag8197 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetaldehyde dehydrogenase 8; aldehyde dehydrogenase 3 family, member B2; Aldehyde dehydrogenase 8; Aldehyde dehydrogenase 8 ALDH3B2 ALDH8; Aldehyde dehydrogenase family 3 member B2
Gene Aliases: AI848594; Aldh3b2; ALDH8; C130048D07Rik
UniProt ID: (Mouse) E9Q3E1
Entrez Gene ID: (Human) 222, (Mouse) 621603, (Rat) 688800
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.