Proteintech
Manufacturer Code:194461AP150UL
Catalog # 50-173-2405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ALDH3B1 Fusion Protein Ag7025 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aldehyde dehydrogenase 3 family, member B1; aldehyde dehydrogenase 3B1; Aldehyde dehydrogenase 7; Aldehyde dehydrogenase 7 ALDH3B1 ALDH4 ALDH7; Aldehyde dehydrogenase family 3 member B1
Gene Aliases: 1700001N19Rik; ALDH3B1; ALDH4; ALDH7
UniProt ID: (Human) P43353, (Mouse) Q80VQ0, (Rat) Q5XI42
Entrez Gene ID: (Human) 221, (Mouse) 67689, (Rat) 309147
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.