Invitrogen
Manufacturer Code:MA525636
Catalog # PIMA525636
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2D3 |
Immunogen | Full length human recombit protein of ALDH3A2 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: alcohol dehydrogenase family 3, subfamily A2; Aldehyde dehydrogenase 10; aldehyde dehydrogenase 3 family, member A2; aldehyde dehydrogenase 3A2; Aldehyde dehydrogenase 4; Aldehyde dehydrogenase family 3 member A2; aldehyde dehydrogenase family 3, subfamily A2; Aldh3a2 aldehyde dehydrogenase 3 family member A2 Aldh4 FALDH alcohol dehydrogenase family 3 subfamily A2 aldehyde dehydrogenase 3A2 aldehyde dehydrogenase 4 aldehyde dehydrogenase family 3 member A2 aldehyde dehydrogenase family 3 subfamily A2 fatty aldehyde dehydrogenase microsomal aldehyde dehydrogenase msALDH; Fatty aldehyde dehydrogenase; Microsomal aldehyde dehydrogenase; msALDH
Gene Aliases: ALDH10; ALDH3A2; Aldh4; FALDH; QccE-15682; SLS
UniProt ID: (Cynomolgus monkey) Q60HH8, (Human) P51648, (Rat) P30839
Entrez Gene ID: (Dog) 100688308, (Cynomolgus monkey) 102138923, (Human) 224, (Rat) 65183
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.