OriGene
Manufacturer Code:CF503143
Catalog # 50-166-1917
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1H10 |
Immunogen | Full length human recombit protein of human ALDH3A2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Aldehyde dehydrogenase 10; aldehyde dehydrogenase 3 family, member A2; Aldehyde dehydrogenase family 3 member A2; ALDH10 FALDH SLS; Fatty aldehyde dehydrogenase; Microsomal aldehyde dehydrogenase
Gene Aliases: ALDH10; ALDH3A2; FALDH; SLS
UniProt ID: (Human) P51648
Entrez Gene ID: (Human) 224
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.