Invitrogen
Manufacturer Code:PA585288
Catalog # PIPA585288
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human ALDH3A2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aldehyde dehydrogenase 10; aldehyde dehydrogenase 3 family, member A2; Aldehyde dehydrogenase family 3 member A2; ALDH10 antibody DKFZp686E23276 antibody FALDH antibody FLJ20851 antibody SLS antibody ALDH3A2 antibody microsomal aldehyde dehydrogenase antibody fatty aldehyde dehydrogenase antibody aldehyde dehydrogenase family 3 member A2 antibody aldehyde dehydrogenase 10 antibody aldehyde dehydrogenase 3 family member A2 antibody; Fatty aldehyde dehydrogenase; Microsomal aldehyde dehydrogenase
Gene Aliases: ALDH10; ALDH3A2; FALDH; SLS
UniProt ID: (Human) P51648
Entrez Gene ID: (Human) 224
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.