Invitrogen
Manufacturer Code:PA569297
Catalog # PIPA569297
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human ALDH3A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alcohol dehydrogenase family 3, subfamily A2; Aldehyde dehydrogenase 10; Aldehyde dehydrogenase 3; aldehyde dehydrogenase 3 family, member A2; aldehyde dehydrogenase 3A2; Aldehyde dehydrogenase 4; Aldehyde dehydrogenase family 3 member A2; aldehyde dehydrogenase family 3, subfamily A2; Aldh3a2 aldehyde dehydrogenase 3 family member A2 Aldh4 FALDH alcohol dehydrogenase family 3 subfamily A2 aldehyde dehydrogenase 3A2 aldehyde dehydrogenase 4 aldehyde dehydrogenase family 3 member A2 aldehyde dehydrogenase family 3 subfamily A2 fatty aldehyde dehydrogenase microsomal aldehyde dehydrogenase msALDH; Fatty aldehyde dehydrogenase; Fatty aldehyde dehydrogenase-like protein; Microsomal aldehyde dehydrogenase; msALDH
Gene Aliases: Ahd-3; Ahd-3r; Ahd3; Ahd3-r; AI194803; ALDH10; Aldh3; ALDH3A2; Aldh4; Aldh4-r; FALDH; SLS
UniProt ID: (Human) P51648, (Rat) P30839, (Mouse) P47740
Entrez Gene ID: (Pig) 100525303, (Rabbit) 100338784, (Dog) 100688308, (Bovine) 513967, (Horse) 100073165, (Human) 224, (Guinea pig) 100728950, (Rat) 65183, (Mouse) 11671
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.