Invitrogen
Manufacturer Code:MA525085
Catalog # PIMA525085
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B6 |
Immunogen | Full length human recombit protein of ALDH3A1 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Aldehyde dehydrogenase 3; aldehyde dehydrogenase 3 family, member A1; Aldehyde dehydrogenase family 3 member A1; aldehyde dehydrogenase isozyme 3; aldehyde dehydrogenase type III; Aldehyde dehydrogenase, dimeric NADP-preferring; ALDH3A1 aldehyde dehydrogenase 3 family memberA1 ALDH3 ALDHIII MGC10406 aldehyde dehydrogenase 3A1 aldehyde dehydrogenase isozyme 3 aldehyde dehydrogenase type III aldehyde dehydrogenase dimeric NADP-preferring stomach aldehyde dehydrogenase; ALDHIII; stomach aldehyde dehydrogenase
Gene Aliases: ALDH3; ALDH3A1; ALDHIII
UniProt ID: (Human) P30838
Entrez Gene ID: (Human) 218
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.