Invitrogen
Manufacturer Code:PA596349
Catalog # PIPA596349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-300 of human ALDH3A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alcohol dehydrogenase family 3, subfamily A1; Aldehyde dehydrogenase 3; aldehyde dehydrogenase 3 family, member A1; aldehyde dehydrogenase 3, stomach cytosolic (class 3); Aldehyde dehydrogenase 4; Aldehyde dehydrogenase family 3 member A1; aldehyde dehydrogenase isozyme 3; aldehyde dehydrogenase type III; Aldehyde dehydrogenase, dimeric NADP-preferring; ALDHIII; Dioxin-inducible aldehyde dehydrogenase 3; stomach aldehyde dehydrogenase
Gene Aliases: Ahd-4; Ahd4; Aldh; ALDH3; ALDH3A1; Aldh4; ALDHIII
UniProt ID: (Human) P30838, (Mouse) P47739
Entrez Gene ID: (Human) 218, (Mouse) 11670
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.