Invitrogen
Manufacturer Code:PA585334
Catalog # PIPA585334
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Bovine |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human aldehyde dehydrogenase 3A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aldehyde dehydrogenase 3 family, memberA1; aldehyde dehydrogenase family 3 member A1; ALDH3 antibody ALDHIII antibody MGC10406 antibody ALDH3A1 antibody aldehyde dehydrogenase dimeric NADP-preferring antibody stomach aldehyde dehydrogenase antibody aldehyde dehydrogenase type III antibody aldehyde dehydrogenase isozyme 3 antibody aldehyde dehydrogenase 3 family member A1 antibody; BCP54; corneal 15.8 kDa protein; corneal protein 54, transparentin; dehydrogenase 3, dimeric NAPD-prefering; transparentin
Gene Aliases: ALDH3
Entrez Gene ID: (Bovine) 281617
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.