Novus Biologicals
Manufacturer Code:NBP267132
Catalog # NBP267132
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | E4-D10 |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: acetaldehyde dehydrogenase 2; acetaldehyde dehydrogenase 2 aldehyde dehydrogenase 2 family (mitochondrial) aldehyde dehydrogenase mitochondrial ALDH class 2 ALDH-E2 ALDHI ALDM EC 1.2.1 EC 1.2.1.3 liver mitochondrial ALDH MGC1806 nucleus-encoded mitochondrial aldehyde dehydrogenase 2; Aldehyde dehydrogenase, mitochondrial; ALDH class 2; ALDH-E2; ALDHI; liver mitochondrial ALDH; nucleus-encoded mitochondrial aldehyde dehydrogenase 2
Gene Aliases: ALDH-E2; ALDH2; ALDHI; ALDM
UniProt ID: (Human) P05091
Entrez Gene ID: (Human) 217
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.