Invitrogen
Manufacturer Code:PA527414
Catalog # PIPA527414
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 151 and 517 of Human ALDH2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetaldehyde dehydrogenase 2; AHD-M1; Aldehyde dehydrogenase, mitochondrial; ALDH class 2; ALDH-E2; ALDH-E2 ALDHI ALDM ALDH class 2 acetaldehyde dehydrogenase 2 aldehyde dehydrogenase mitochondrial liver mitochondrial ALDH nucleus-encoded mitochondrial aldehyde dehydrogenase 2; ALDHI; liver mitochondrial ALDH; nucleus-encoded mitochondrial aldehyde dehydrogenase 2
Gene Aliases: Ahd-1; Ahd-5; AHD-M1; Ahd1; Ahd5; ALDH-E2; ALDH2; ALDHI; ALDM
UniProt ID: (Human) P05091, (Mouse) P47738
Entrez Gene ID: (Human) 217, (Mouse) 11669
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.