OriGene
Manufacturer Code:UM500040CF
Catalog # 50-168-4914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | UMAB44 |
Immunogen | Full length human recombit protein of human ALDH1L1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 10-formyltetrahydrofolate dehydrogenase; 10-FTHFDH; 10-FTHFDH 10-fTHF FDH FTHFD; aldehyde dehydrogenase 1 family, member L1; Aldehyde dehydrogenase family 1 member L1; Cytosolic 10-formyltetrahydrofolate dehydrogenase; MER57B-ALDH1L1
Gene Aliases: 10-fTHF; 10-FTHFDH; ALDH1L1; FDH; FTHFD
UniProt ID: (Human) O75891
Entrez Gene ID: (Human) 10840, (Dog) 100855730
Molecular Function:
dehydrogenase
oxidoreductase
methyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.