Invitrogen
Manufacturer Code:PA595376
Catalog # PIPA595376
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human ALDH1B1 different from the related mouse sequence by one amino acid and from the related rat sequence by two amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetaldehyde dehydrogenase 5; aldehyde dehydrogenase 1 family, member B1; aldehyde dehydrogenase 1B1; Aldehyde dehydrogenase 5; Aldehyde dehydrogenase family 1 member B1; Aldehyde dehydrogenase X mitochondrial 1.2.1.3 Aldehyde dehydrogenase 5 Aldehyde dehydrogenase family 1 member B1 ALDH1B1 ALDH5 ALDHX; Aldehyde dehydrogenase X, mitochondrial; ALDH class 2
Gene Aliases: 2700007F14Rik; ALDH1B1; ALDH5; ALDHX
UniProt ID: (Human) P30837, (Mouse) Q9CZS1, (Rat) Q66HF8
Entrez Gene ID: (Human) 219, (Mouse) 72535, (Rat) 298079
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.