Invitrogen
Manufacturer Code:PA519327
Catalog # PIPA519327
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DKEQFERVLGYIQ) corresponding to the internal amino acids of ALDH1B1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: acetaldehyde dehydrogenase 5; aldehyde dehydrogenase 1 family, member B1; Aldehyde dehydrogenase 5; Aldehyde dehydrogenase family 1 member B1; Aldehyde dehydrogenase X, mitochondrial; ALDH class 2; ALDH1B1 aldehyde dehydrogenase 1 family member B1 ALDH5 ALDHX MGC2230 ALDH class 2 acetaldehyde dehydrogenase 5 aldehyde dehydrogenase 1B1 aldehyde dehydrogenase 5 mitochondrial aldehyde dehydrogenase X
Gene Aliases: ALDH1B1; ALDH5; ALDHX
UniProt ID: (Human) P30837
Entrez Gene ID: (Human) 219
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.