Invitrogen
Manufacturer Code:PA5109378
Catalog # PIPA5109378
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 278-517 of human ALDH1B1 (NP_000683.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2700007F14Rik Aldehyde dehydrogenase 5 Aldehyde dehydrogenase family 1 member B1 Aldehyde dehydrogenase X mitochondrial Aldh1b1 ALDH5 Aldhx MGC2230; acetaldehyde dehydrogenase 5; aldehyde dehydrogenase 1 family, member B1; aldehyde dehydrogenase 1B1; Aldehyde dehydrogenase 5; Aldehyde dehydrogenase family 1 member B1; Aldehyde dehydrogenase X, mitochondrial; ALDH class 2
Gene Aliases: 2700007F14Rik; ALDH1B1; ALDH5; ALDHX
UniProt ID: (Human) P30837, (Mouse) Q9CZS1, (Rat) Q66HF8
Entrez Gene ID: (Human) 219, (Mouse) 72535, (Rat) 298079
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.