Invitrogen
Manufacturer Code:PA515000
Catalog # PIPA515000
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 24-51 amino acids from the N-terminal region of human ALDH1A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetaldehyde dehydrogenase 6; aldehyde dehydrogenase 1 family, member A3; Aldehyde dehydrogenase 6; Aldehyde dehydrogenase family 1 member A3; aldehyde dehydrogenase-6; RALDH-3; Retinaldehyde dehydrogenase 3; Retinaldehyde dehydrogenase 3 Aldehyde dehydrogenase 6 Aldehyde dehydrogenase family 1 member A3 ALDH1A3 ALDH6 ALDHA1A3
Gene Aliases: ALDH1A3; ALDH1A6; ALDH6; MCOP8; RALDH3; V1
UniProt ID: (Human) P47895, (Mouse) Q9JHW9
Entrez Gene ID: (Human) 220, (Mouse) 56847
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.