Invitrogen
Manufacturer Code:PA552684
Catalog # PIPA552684
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human ALDH1A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AL1A2 aldehyde dehydrogenase 1 family member A2 aldehyde dehydrogenase 1A2 aldehyde dehydrogenase family 1 member A2; alcohol dehydrogenase family 1, subfamily A2; alcohol dehydrogenase family 1, subfamily A7; aldehyde dehydrogenase 1 family, member A2; Aldehyde dehydrogenase family 1 member A2; RALDH 2; RALDH(II); Retinal dehydrogenase 2; retinaldehyde dehydrogenase 2; Retinaldehyde-specific dehydrogenase type 2
Gene Aliases: ALDH1A2; Aldh1a7; AV116159; RALDH(II); Raldh1; RALDH2; RALDH2-T
UniProt ID: (Human) O94788, (Mouse) Q62148
Entrez Gene ID: (Human) 8854, (Mouse) 19378
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.