Novus Biologicals
Manufacturer Code:NBP215338
Catalog # NBP215338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human ALDH1A2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aldehyde dehydrogenase 1 family member A2 Aldehyde dehydrogenase family 1 member A2 EC 1.2.1 EC 1.2.1.36 RALDH 2 RALDH(II) RalDH2 RALDH2MGC26444 RALDH2-T retinal dehydrogenase 2 Retinaldehyde-specific dehydrogenase type 2; aldehyde dehydrogenase 1 family, member A2; Aldehyde dehydrogenase family 1 member A2; RALDH 2; RALDH(II); Retinal dehydrogenase 2; Retinaldehyde-specific dehydrogenase type 2
Gene Aliases: ALDH1A2; RALDH(II); RALDH2; RALDH2-T
UniProt ID: (Human) O94788
Entrez Gene ID: (Human) 8854
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.