Invitrogen
Manufacturer Code:PA532159
Catalog # PIPA532159
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 232 of Human ALDH1A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetaldehyde dehydrogenase 1; ALDC ALDH-E1 ALDH1 ALDH11 PUMB1 RALDH1 ALDH class 1 ALHDII RALDH 1 acetaldehyde dehydrogenase 1 aldehyde dehydrogenase 1 soluble aldehyde dehydrogenase liver cytosolic retinal dehydrogenase 1 retinaldehyde dehydrogenase 1; aldehyde dehydrogenase 1 family, member A1; aldehyde dehydrogenase 1, soluble; Aldehyde dehydrogenase family 1 member A1; Aldehyde dehydrogenase, cytosolic; aldehyde dehydrogenase, liver cytosolic; ALDH class 1; ALDH-E1; ALHDII; epididymis luminal protein 12; epididymis luminal protein 9; epididymis secretory sperm binding protein Li 53e; RALDH 1; Retinal dehydrogenase 1; retinaldehyde dehydrogenase 1
Gene Aliases: ALDC; ALDH-E1; ALDH1; ALDH11; ALDH1A1; HEL-9; HEL-S-53e; HEL12; PUMB1; RALDH1
UniProt ID: (Human) P00352
Entrez Gene ID: (Human) 216
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.