Proteintech
Manufacturer Code:219721AP150UL
Catalog # 50-173-3664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ALCAM Fusion Protein Ag16892 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Activated leukocyte cell adhesion molecule; ALCAM CD166 CD166 antigen MEMD; CD166; CD166 antigen
Gene Aliases: ALCAM; CD166; MEMD
UniProt ID: (Human) Q13740
Entrez Gene ID: (Human) 214
Molecular Function:
cell adhesion molecule
immunoglobulin superfamily cell adhesion molecule
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.