Invitrogen
Manufacturer Code:PA521553
Catalog # PIPA521553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 211 and 587 of ALAS-E (Uniprot IDNo.P22557) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5-aminolevulinate synthase, erythroid-specific, mitochondrial; 5-aminolevulinic acid synthase 2; ALAS-E; aminolevulinate, delta-, synthase 2; Delta-ALA synthase 2; delta-ALA synthetase; Delta-aminolevulinate synthase 2; erythroid-specific ALAS; RP5-884M20.1 ALAS-E ALASE ANH1 ASB XLDPP XLEPP XLSA 5-aminolevulinate synthase erythroid-specific mitochondrial 5-aminolevulinic acid synthase 2 delta-ALA synthase 2 delta-ALA synthetase delta-aminolevulinate synthase 2
Gene Aliases: ALAS; Alas-2; ALAS-E; ALAS2; ALASE; ANH1; ASB; SIDBA1; XLDPP; XLEPP; XLSA
UniProt ID: (Human) P22557, (Mouse) P08680
Entrez Gene ID: (Human) 212, (Mouse) 11656
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.