Novus Biologicals
Manufacturer Code:NBP201724
Catalog # NBP201724
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4D6 |
Immunogen | This AKT1 antibody is made to the full length human recombinant protein of human AKT1(NP_005154) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AKT AKT serine/threonine kinase 1 AKT1m CWS6 EC 2.7.11 EC 2.7.11.1 PKB PKB alpha PKBMGC99656 PRKBA Protein kinase B Protein Kinase B Alpha Proto-oncogene c-Akt RAC rac protein kinase alpha RAC-ALPHA RAC-alpha serine/threonine-protein kinas; AKT1m; PKB; PKB alpha; Protein kinase B; Protein kinase B alpha; Proto-oncogene c-Akt; rac protein kinase alpha; RAC-alpha serine/threonine-protein kinase; RAC-PK-alpha; serine-threonine protein kinase; v-akt murine thymoma viral oncogene homolog 1; v-akt murine thymoma viral oncogene-like protein 1
Gene Aliases: AKT; AKT1; CWS6; PKB; PKB-ALPHA; PRKBA; RAC; RAC-ALPHA
UniProt ID: (Human) P31749
Entrez Gene ID: (Human) 207
Molecular Function: annexin calcium-binding protein calmodulin intracellular calcium-sensing protein kinase non-receptor serine/threonine protein kinase protein kinase transfer/carrier protein transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.