Invitrogen
Manufacturer Code:PA513999
Catalog # PIPA513999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 101-129 amino acids from the N-terminal region of human AKR7A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AFB1 aldehyde reductase 1; AFB1-AR 1; aflatoxin aldehyde reductase; Aflatoxin B1 aldehyde reductase member 2; Aflatoxin B1 aldehyde reductase member 2 AFB1-AR 1 Aldoketoreductase 7 synonyms: AKR7 synonyms: AFAR1 synonyms: AFAR name: AKR7A2; aflatoxin beta1 aldehyde reductase; akr7a2; aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase); Aldoketoreductase 7; epididymis secretory sperm binding protein Li 166mP; HEL-S-166mP; SSA reductase; Succinic semialdehyde reductase
Gene Aliases: 0610025K21Rik; AFAR; AFAR1; AFB1-AR1; AKR7; AKR7A2; Akr7a5
UniProt ID: (Human) O43488, (Mouse) Q8CG76
Entrez Gene ID: (Human) 8574, (Mouse) 110198
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.