Invitrogen
Manufacturer Code:PA5102398
Catalog # PIPA5102398
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AKR7A2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610025K21Rik Afar AFAR1 Afar2 AFB1 aldehyde reductase 1 AFB1-AR 1 AFB1-AR1 aflatoxin aldehyde reductase aflatoxin B1 aldehyde reductase member 2 aflatoxin beta1 aldehyde reductase Aiar AKR7 AKR7A2 Akr7a5 aldo-keto reductase family 7 member A2 aldo-keto reductase family 7 member A2 (aflatoxin aldehyde reductase) aldo-keto reductase family 7 member A5 (aflatoxin aldehyde reductase) Aldoketoreductase 7 epididymis secretory sperm binding protein Li 166mP HEL-S-166mP rAFAR2 SSA reductase succinic semialdehyde reductase synonyms: AFAR1; AFB1 aldehyde reductase 1; AFB1-AR 1; aflatoxin aldehyde reductase; Aflatoxin B1 aldehyde reductase member 2; aflatoxin beta1 aldehyde reductase; akr7a2; aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase); Aldoketoreductase 7; epididymis secretory sperm binding protein Li 166mP; HEL-S-166mP; rAFAR2; SSA reductase; Succinic semialdehyde reductase
Gene Aliases: 0610025K21Rik; AFAR; AFAR1; Afar2; AFB1-AR1; Aiar; AKR7; AKR7A2; Akr7a5
UniProt ID: (Human) O43488, (Rat) Q8CG45, (Mouse) Q8CG76
Entrez Gene ID: (Human) 8574, (Rat) 171445, (Mouse) 110198
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.