Abnova
Manufacturer Code:H00008574K
Catalog # 89-209-728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human AKR7A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human AKR7A2 peptide using ARM Technology.
For Research Use Only
Protein Aliases: AFAR AFAR1 AFB1-AR1 AKR7; AFB1 aldehyde reductase 1; AFB1-AR 1; aflatoxin aldehyde reductase; Aflatoxin B1 aldehyde reductase member 2; aflatoxin beta1 aldehyde reductase; aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase); Aldoketoreductase 7; epididymis secretory sperm binding protein Li 166mP; HEL-S-166mP; SSA reductase; Succinic semialdehyde reductase
Gene Aliases: AFAR; AFAR1; AFB1-AR1; AKR7; AKR7A2
UniProt ID: (Human) O43488
Entrez Gene ID: (Human) 8574
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.