Invitrogen
Manufacturer Code:MA535413
Catalog # PIMA535413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0857 |
Immunogen | A synthesized peptide derived from human AKR1C3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 5; 17-beta-HSD 5 17beta-HSD5 17-beta-hydroxysteroid dehydrogenase type 5 20-alpha-HSD 20-alpha-hydroxysteroid dehydrogenase 2-dih 3-alpha-HSD type 2 3-alpha-HSD type II brain 3-alpha-hydroxysteroid dehydrogenase type 2 Akr1c18 AKR1C3 Aldo-keto reductase family 1 member C18 Aldo-keto reductase family 1 member C3 Chlordecone reductase homolog HAKRb DD3 DD-3 DDH1 DDX Dihydrodiol dehydrogenase 3 Dihydrodiol dehydrogenase type I HA1753 HAKRB HAKRe hluPGFS HSD1 HSD17B5 Indanol dehydrogenase KIAA0119 PGFS Prostaglandin F synthase Testosterone 17-beta-dehydrogenase 5 Trans-12-dihydrobenzene-12-diol dehydrogenase; 17-beta-hydroxysteroid dehydrogenase type 5; 3-alpha hydroxysteroid dehydrogenase, type II; 3-alpha-HSD type 2; 3-alpha-HSD type II, brain; 3-alpha-hydroxysteroid dehydrogenase type 2; Aldo-keto reductase family 1 member C3; Chlordecone reductase homolog HAKRb; DD-3; Dihydrodiol dehydrogenase 3; Dihydrodiol dehydrogenase type I; dihydrodiol dehydrogenase X; HA1753; indanol dehydrogenase; PGFS; Prostaglandin F synthase; Testosterone 17-beta-dehydrogenase 5; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; type IIb 3-alpha hydroxysteroid dehydrogenase
Gene Aliases: AKR1C3; DD3; DDH1; DDX; HA1753; HAKRB; HAKRe; hluPGFS; HSD17B5; KIAA0119; PGFS
UniProt ID: (Human) P42330
Entrez Gene ID: (Human) 8644
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.