Invitrogen
Manufacturer Code:PA518339
Catalog # PIPA518339
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CFASHPNYPYSDEY) corresponding to the C-terminus amino acids of AKR1C3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 5; 17-beta-hydroxysteroid dehydrogenase type 5; 3-alpha hydroxysteroid dehydrogenase, type II; 3-alpha-HSD type 2; 3-alpha-HSD type II, brain; 3-alpha-hydroxysteroid dehydrogenase type 2; AKR1C3 DD3 HAKRB HAKRe HA1753 HSD17B5 hluPGFS KIAA0119 aldo-keto reductase family 1 member C3 (3-alpha hydroxysteroid dehydrogenase type II) prostaglandin F synthase dihydrodiol dehydrogenase 3 chlordecone reductase homolog hydroxysteroid (17-beta) dehydrogenase 5 type IIb 3-alpha hydroxysteroid dehydrogenase trans-12-dihydrobenzene-12-diol dehydrogenase DDX aldo-keto reductase family 1 member C3 chlordecone reductase dihydrodiol dehydrogenase X type II 3a-hydroxysteroid; Aldo-keto reductase family 1 member C3; Chlordecone reductase homolog HAKRb; DD-3; Dihydrodiol dehydrogenase 3; Dihydrodiol dehydrogenase type I; dihydrodiol dehydrogenase X; HA1753; indanol dehydrogenase; PGFS; Prostaglandin F synthase; Testosterone 17-beta-dehydrogenase 5; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; type IIb 3-alpha hydroxysteroid dehydrogenase
Gene Aliases: AKR1C3; DD3; DDH1; DDX; HA1753; HAKRB; HAKRe; hluPGFS; HSD17B5; KIAA0119; PGFS
UniProt ID: (Human) P42330
Entrez Gene ID: (Human) 8644
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.