Invitrogen
Manufacturer Code:PA5101414
Catalog # PIPA5101414
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AKR1C2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-alpha-HSD 17-alpha-hydroxysteroid dehydrogenase 3 alpha-HSD3 3(17)alpha-hydroxysteroid dehydrogenase 3(or 17)-alpha-hydroxysteroid dehydrogenase 3-alpha-HSD3 3-alpha-hydroxysteroid dehydrogenase 9430025F20Rik AI315367 AKR1C2 Akr1c21 AKR1C-pseudo Aldo-keto reductase family 1 member C2 aldo-keto reductase family 1 member C21 aldo-keto reductase family 1 member C2 aldo-keto reductase family 1 member C21 BABP Chlordecone reductase homolog HAKRD DD DD/BABP DD1 DD2 DD-2 DD3 DDH2 Dihydrodiol dehydrogenase 2 dihydrodiol dehydrogenase 2 bile acid binding protein 3-alpha hydroxysteroid dehydrogenase type III dihydrodiol dehydrogenase type 1 Dihydrodiol dehydrogenase type 3 Dihydrodiol dehydrogenase/bile acid-binding protein HAKRD HBAB HSD3-3 alpha HSD3-alpha3 MCDR2 pseudo-chlordecone reductase SRXY8 TDD testicular 1720-desmolase deficiency trans-12-dihydrobenzene-12-diol dehydrogenase type II dihydrodiol dehydrogenase Type III 3-alpha-hydroxysteroid dehydrogenase; 3-alpha-HSD3; Aldo-keto reductase family 1 member C2; Chlordecone reductase homolog HAKRD; DD-2; DD/BABP; Dihydrodiol dehydrogenase 2; dihydrodiol dehydrogenase 2; bile acid binding protein; 3-alpha hydroxysteroid dehydrogenase, type III; Dihydrodiol dehydrogenase/bile acid-binding protein; pseudo-chlordecone reductase; testicular 17,20-desmolase deficiency; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; type II dihydrodiol dehydrogenase; Type III 3-alpha-hydroxysteroid dehydrogenase
Gene Aliases: AKR1C-pseudo; AKR1C2; BABP; DD; DD-2; DD/BABP; DD2; DDH2; HAKRD; HBAB; MCDR2; SRXY8; TDD
UniProt ID: (Human) P52895
Entrez Gene ID: (Human) 1646
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.