Invitrogen
Manufacturer Code:PA5114836
Catalog # PIPA5114836
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human AKR1C1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-beta-HSD 5 17HSD type 5 20 alpha-hydroxysteroid dehydrogenase 20-alpha (3-alpha)-hydroxysteroid dehydrogenase) 20-ALPHA-HSD 20-alpha-hydroxysteroid dehydrogenase 2-ALPHA-HSD 3alpha-HSD Akr1c1 Akr1c6 aldo-keto reductase C aldo-keto reductase family 1 member C1 aldo-keto reductase family 1 member C1 aldo-keto reductase family 1 member C1 (aldehyde reductase) aldo-keto reductase family 1 member C1 (dihydrodiol dehydrogenase 1 aldo-keto reductase family 1 member C1 (dihydrodiol dehydrogenase 1 20-alpha (3-alpha)-hydroxysteroid dehydrogenase) aldo-keto reductase family 1 member C6 C9 Chlordecone reductase homolog HAKRC DD1 DD1/DD2 DDH DDH1 dihydrodiol dehydrogenase 1 dihydrodiol dehydrogenase 1/2 dihydrodiol dehydrogenase 1 20-alpha (3-alpha)-hydroxysteroid dehydrogenase dihydrodiol dehydrogenase isoform DD1 estradiol 17 beta-dehydrogenase 5 estradiol 17-beta-dehydrogenase (A-specific) H-37 HAKRC HBAB hepatic dihydrodiol dehydrogenase High-affinity hepatic bile acid-binding protein Hsd17b5 hydroxysteroid 17-beta dehydrogenase 5 Indanol dehydrogenase MBAB RP11-69D4.1 Trans-12-dihydrobenzene-12-diol dehydrogenase type 5 17beta-hydroxysteroid dehydrogenase type II 3-alpha-hydroxysteroid dehydrogenase; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; 20-alpha-hydroxysteroid dehydrogenase; aldo-keto reductase C; Aldo-keto reductase family 1 member C1; Chlordecone reductase homolog HAKRC; DD1; Dihydrodiol dehydrogenase 1; dihydrodiol dehydrogenase 1/2; dihydrodiol dehydrogenase 1; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase; dihydrodiol dehydrogenase isoform DD1; HBAB; hepatic dihydrodiol dehydrogenase; High-affinity hepatic bile acid-binding protein; indanol dehydrogenase; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; type II 3-alpha-hydroxysteroid dehydrogenase
Gene Aliases: 2-ALPHA-HSD; 20-ALPHA-HSD; AKR1C1; C9; DD1; DD1/DD2; DDH; DDH1; H-37; HAKRC; HBAB; MBAB
UniProt ID: (Human) Q04828
Entrez Gene ID: (Human) 1645
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.