Invitrogen
Manufacturer Code:PA578752
Catalog # PIPA578752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human AKR1C1/C2 recombit protein (Position: M1-K123). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 17-alpha-HSD; 17-alpha-hydroxysteroid dehydrogenase; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase); 20-alpha-HSD; 20-alpha-hydroxysteroid dehydrogenase; 3(or 17)-alpha-hydroxysteroid dehydrogenase; 3-alpha-HSD3; 3-alpha-hydroxysteroid dehydrogenase; aldo-keto reductase C; Aldo-keto reductase family 1 member C1; Aldo-keto reductase family 1 member C2; Aldo-keto reductase family 1 member C2 3-alpha-HSD3 Chlordecone reductase homolog HAKRD Dihydrodiol dehydrogenase 2 DD-2 DD2 Dihydrodiol dehydrogenase/bile acid-binding protein DD/BABP Trans-12-dihydrobenzene-12-diol dehydrogenase; Aldo-keto reductase family 1 member C21; aldo-keto reductase family 1, member C1; aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase); aldo-keto reductase family 1, member C21; aldo-keto reductase family 1, member C6; Chlordecone reductase homolog HAKRC; Chlordecone reductase homolog HAKRD; DD-2; DD/BABP; DD1; Dihydrodiol dehydrogenase 1; dihydrodiol dehydrogenase 1/2; dihydrodiol dehydrogenase 1; 20-alpha (3-alpha)-hydroxysteroid dehydrogenase; Dihydrodiol dehydrogenase 2; dihydrodiol dehydrogenase 2; bile acid binding protein; 3-alpha hydroxysteroid dehydrogenase, type III; dihydrodiol dehydrogenase isoform DD1; Dihydrodiol dehydrogenase/bile acid-binding protein; HBAB; hepatic dihydrodiol dehydrogenase; High-affinity hepatic bile acid-binding protein; indanol dehydrogenase; pseudo-chlordecone reductase; testicular 17,20-desmolase deficiency; trans-1,2-dihydrobenzene-1,2-diol dehydrogenase; type 5 17beta-hydroxysteroid dehydrogenase; type II 3-alpha-hydroxysteroid dehydrogenase; type II dihydrodiol dehydrogenase; Type III 3-alpha-hydroxysteroid dehydrogenase
Gene Aliases: 2-ALPHA-HSD; 20-ALPHA-HSD; AKR1C-pseudo; AKR1C1; AKR1C2; Akr1c21; Akr1c6; BABP; C9; DD; DD-2; DD/BABP; DD1; DD1/DD2; DD2; DDH; DDH1; DDH2; H-37; HAKRC; HAKRD; HBAB; Hsd17b5; MBAB; MCDR2; SRXY8; TDD
UniProt ID: (Human) Q04828, (Human) P52895, (Rat) Q6AYQ2
Entrez Gene ID: (Human) 1645, (Human) 1646, (Rat) 307092, (Rat) 291283
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.