Abnova
Manufacturer Code:MAB9984
Catalog # 89-113-942
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | Recombinant protein corresponding to amino acids 33-316 of human AKR1B1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant AKR1B1.
For Research Use Only
Protein Aliases: ADR; Aldehyde reductase; aldehyde reductase 1; Aldo-keto reductase family 1 member B1; aldo-keto reductase family 1, member B1 (aldose reductase); Aldose reductase; ALDR1; ALR2; AR; Lii5-2 CTCL tumor antigen; low Km aldose reductase; MGC1804
Gene Aliases: ADR; AKR1B1; ALDR1; ALR2; AR
UniProt ID: (Human) P15121
Entrez Gene ID: (Human) 231
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.