Invitrogen
Manufacturer Code:PA578750
Catalog # PIPA578750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human AKR1B1 recombit protein (Position: L228-F316). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aldehyde reductase; aldehyde reductase 1; Aldehyde reductase 1 (low Km aldose reductase) (5.8 kb PstI fragment, probably the functional gene); Aldo-keto reductase family 1 member B1; Aldo-keto reductase family 1 member B3; aldo-keto reductase family 1, member B1 (aldose reductase); aldo-keto reductase family 1, member B4 (aldose reductase); Aldose reductase; Aldose reductase AR Aldehyde reductase Aldo-keto reductase family 1 member B1 AKR1B1 ALDR1; AR; Lii5-2 CTCL tumor antigen; low Km aldose reductase
Gene Aliases: ADR; Ahr-1; Ahr1; AKR1B1; Akr1b3; Akr1b4; Aldor1; ALDR1; ALDRED; Alr; ALR-P-I; ALR2; AR; RATALDRED
UniProt ID: (Human) P15121, (Rat) P07943, (Mouse) P45376
Entrez Gene ID: (Human) 231, (Rat) 24192, (Mouse) 11677
Molecular Function:
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.