Invitrogen
Manufacturer Code:PA518073
Catalog # PIPA518073
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DAGHPLYPFNDPY) corresponding to the C-terminus amino acids of AKR1A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: alcohol dehydrogenase; Alcohol dehydrogenase [NADP(+)]; Aldehyde reductase; aldehyde reductase AKR1A1 aldo-keto reductase family 1 member A1 (aldehyde reductase) ALDR1 ALR MGC12529 MGC1380 alcohol dehydrogenase aldo-keto reductase family 1 member A1 DD3 OTTHUMP00000009240 dihydrodiol dehydrogenase 3; Aldo-keto reductase family 1 member A1; aldo-keto reductase family 1, member A1 (aldehyde reductase); dihydrodiol dehydrogenase 3; epididymis secretory protein Li 6; epididymis secretory sperm binding protein Li 165mP; Glucuronate reductase; Glucuronolactone reductase; HEL-S-165mP
Gene Aliases: AKR1A1; ALDR1; ALR; ARM; DD3; HEL-S-6
UniProt ID: (Human) P14550
Entrez Gene ID: (Human) 10327
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.