Invitrogen
Manufacturer Code:PA559993
Catalog # PIPA559993
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human AKAP8L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A kinase (PRKA) anchor protein 8-like; A-kinase anchor protein 8-like; AKAP8-like protein; HA95; HA95 HAP95 NAKAP NAKAP95 A-kinase anchor protein 8-like AKAP8-like protein helicase A-binding protein 95 kDa homologous to AKAP95 protein neighbor of A kinase anchoring protein 95 neighbor of A-kinase anchoring protein 95 neighbor of A-kinase-anchoring protein 95 neighbor of AKAP; HAP95; Helicase A-binding protein 95; helicase A-binding protein 95 kDa; Homologous to AKAP95 protein; neighbor of A-kinase anchoring protein 95; Neighbor of A-kinase-anchoring protein 95; Neighbor of AKAP95; testis tissue sperm-binding protein Li 90mP
Gene Aliases: AKAP8L; HA95; HAP95; HRIHFB2018; NAKAP; NAKAP95
UniProt ID: (Human) Q9ULX6
Entrez Gene ID: (Human) 26993
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.